Learn More
Fisherbrand™ Single-Channel Pipette
Aspirates and dispenses viscous liquids accurately and reliably. Fisherbrand™ Single-Channel Pipette include a lock counter to prevent accidental volume changes. Lightweight, ergonomic design minimizes repetitive strain injuries.
Brand:  Fisherbrand™ FI8-101-00-9
52.24 EUR valid until 2024-12-20
Use promo code "21644" to get your promotional price.
 View more versions of this product
Product Code. 10676813
  • 172.78 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
Description
Description
  • Accurate and reliable pipetting
  • Reverse pipetting option for accurate and precise aspirating and dispensing of viscous liquids
  • Innovative soft grip prevents transfer of heat to internal components, ensuring accurate pipette operation
  • Lock counter mechanism of variable volume pipettes prevents volume changes while pipetting
  • Comfortable and effortless pipetting - light weight / ergonomic design for easy operation prevents hand stress. Ergonomic low plunger force minimizes RSI (repetitive strain injury)
  • Pipette is fully autoclavable for sterile handling
  • Easy and quick maintenance and adjustment with the included disassembly and calibration tool
  • No loss of sample - blow out system allows last drop dispensing
  • Meets following standards: DIN EN ISO 8655-1 | DIN EN ISO 8655-2 | DIN EN ISO 8655-6 | EU Directive 98/79/EG
Specifications
Specifications
Autoclavable
Sterile
0.5 to 10 μL
0.01μL
2 Years
1
Manual Pipette
Lock counter mechanism, disassembly and calibration tool, sample bag with tips, grease
1 Each