Learn More
Siponimod, MedChemExpress
Product Code. 16405846
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
500mg
50mg
5mg
Product Code. 16405846
Brand: Medchem Express HY12355
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Siponimod (BAF-312) is an orally active and selective sphingosine-1-phosphate (S1P) receptor modulator. Siponimod is selective for S1P1 and S1P5 over S1P2, S1P3, and S1P4, with EC50s of 0.4, 0.98, >10000, >1000, and 750 nM, respectively. Siponimod can be used for multiple sclerosis (MS) research-.
Siponimod (BAF-312) is an orally active and selective sphingosine-1-phosphate (S1P) receptor modulator. Siponimod is selective for S1P1 and S1P5 over S1P2, S1P3, and S1P4, with EC50s of 0.4, 0.98, >10000, >1000, and 750 nM, respectively. Siponimod can be used for multiple sclerosis (MS) research-.
Specifications
Chemical Name or Material Siponimod
CAS 1230487-00-9
Color White
Purity Grade Notes Research
Molecular Formula C29H35F3N2O3
Quantity 100 mg
Synonym BAF-312
Solubility Information DMSO : ≥ 30 mg/mL (58.07 mM)
SMILES O=C(C1CN(CC2=CC=C(/C(C)=N/OCC3=CC=C(C4CCCCC4)C(C(F)(F)F)=C3)C=C2CC)C1)O
Molecular Weight (g/mol) 516.6
Formula Weight 516.6
Percent Purity 98.0%
Grade Research
Physical Form Solid