Learn More
Siponimod, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
500mg
50mg
5mg
Description
Siponimod (BAF-312) is an orally active and selective sphingosine-1-phosphate (S1P) receptor modulator. Siponimod is selective for S1P1 and S1P5 over S1P2, S1P3, and S1P4, with EC50s of 0.4, 0.98, >10000, >1000, and 750 nM, respectively. Siponimod can be used for multiple sclerosis (MS) research-.
Specifications
Specifications
| Chemical Name or Material | Siponimod |
| CAS | 1230487-00-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C29H35F3N2O3 |
| Quantity | 1 g |
| Synonym | BAF-312 |
| Solubility Information | DMSO : ≥ 30 mg/mL (58.07 mM) |
| SMILES | O=C(C1CN(CC2=CC=C(/C(C)=N/OCC3=CC=C(C4CCCCC4)C(C(F)(F)F)=C3)C=C2CC)C1)O |
| Molecular Weight (g/mol) | 516.6 |
| Show More |