missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC1A5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Bio-Techne NBP1-89327
This item is not returnable.
View return policy
Description
SLC1A5 Polyclonal specifically detects SLC1A5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SLC1A5 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Q15758 | |
SLC1A5 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:VDRTESRSTEPELIQVKSELPLDPLPVPTEEGNPLLKHYRGPAGDATVASEKESVM | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
AAAT, ASCT2M7VS1, ATB(0), Baboon M7 virus receptor, M7V1ATBO, neutral amino acid transporter B, neutral amino acid transporter B(0), R16, RD114 virus receptor, RD114/simian type D retrovirus receptor, RDR, RDRCFLJ31068, Sodium-dependent neutral amino acid transporter type 2, solute carrier family 1 (neutral amino acid transporter), member 5, Solute carrier family 1 member 5 | |
Rabbit | |
Affinity Purified | |
RUO | |
6510 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction