Learn More
Ibidi μ-Slide 8 Well Glass Bottom
μ-Slide 8 Well Glass Bottom is a chambered coverslip with 8 wells.
Brand:  Ibidi 80827
Encompass_Preferred
Product Code. 17226213
  • 268.62 € / Pack of 15
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • A chambered coverslip with 8 wells for use in TIRF and super-resolution microscopy
  • bottom material: #1.5H (170 μm ± 5 μm) D 263 M Schott glass coverslip
  • sterilized
Specifications
Specifications
Chambered Coverglass
8
Sterile