Learn More
Ibidi μ-Slide 8 Well ibiTreat
μ-Slide 8 Well ibiTreat is a chambered coverslip with 8 wells.
Brand:  Ibidi 80826
Encompass_Preferred
Product Code. 15588389
  • 199.65 € / Pack of 15
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • A chambered coverslip with 8 wells for cell culture, immunofluorescence, and high-end microscopy
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • sterilized
Specifications
Specifications
Chambered Coverglass
8