Learn More
ibidi μ-Slide Angiogenesis ibiTreat
A chambered coverslip with 15 wells
Brand:  ibidi 81506
Encompass_Preferred
Product Code. 17296223
  • 321.86 € / Pack of 15
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • A chambered coverslip with 15 wells
  • For tube formation assays, 3D cell culture, and immunofluorescence
  • Bottom material: #1.5 Polymer Coverslip
  • Tissue culture treated
  • Sterilized
Specifications
Specifications
15
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers