Learn More
ibidi μ-Slide Angiogenesis ibiTreat
A chambered coverslip with 15 wells
Brand: ibidi 81506
This item is not returnable.
View return policy
Description
- A chambered coverslip with 15 wells
- For tube formation assays, 3D cell culture, and immunofluorescence
- Bottom material: #1.5 Polymer Coverslip
- Tissue culture treated
- Sterilized
Specifications
15 |