Learn More
Ibidi μ-Slide Chemotaxis ibiTreat
μ-Slide Chemotaxis ibiTreat is a ready-to-use microscopy slide for chemotaxis assays.
Brand:  Ibidi 80326
Encompass_Preferred
Product Code. 13478749
  • 457.38 € / Pack of 10
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • A ready-to-use microscopy slide for chemotaxis assays with fast or slow migrating cells
  • suitable for 2D or 3D cell culture
  • stable chemotactic gradients for long-term experiments
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • sterilized
Specifications
Specifications
Microscope Slide
10