Learn More
Ibidi μ-Slide I ibiTreat
A μ-Slide has one channel for imaging inside a channel.
Brand: Ibidi 80106
This item is not returnable.
View return policy
Description
- A combination of a cell culture chamber and a coverslip for imaging inside a channel
- bottom material: #1.5 Polymer Coverslip
- tissue culture treated
- sterilized
Specifications
Microscope Slide | |
15 |