Learn More
Ibidi μ-Slide I Luer Variety Pack ibiTreat
μ-Slide I Luer Variety Pack ibiTreat includes a variety of μ-Slide I Luer channel slides.
Brand:  Ibidi 81106
Encompass_Preferred
Product Code. 17276213
  • 271.04 € / Pack of 16
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Variety of μ-Slide I Luer channel slides with 4 times each channel height
  • specially suited for optimizing experimental conditions in flow applications
  • easy to connect with tubing and pump systems via female Luers
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • sterilized
Specifications
Specifications
Microscope Slide
16