Learn More
Ibidi μ-Slide VI 0.4 μ-Pattern RGD, sqr30, pit110, hex Trial Pack
μ-Slide VI 0.4 μ-Pattern RGD, sqr30, pit110, hex Trial Pack is a 6 channel slide with micropatterned surface.
Brand:  Ibidi 83603-S
Encompass_Preferred
Product Code. 17824784
  • 174.24 € / Pack of 2
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • A 6 channel slide with micropatterned surface for multi-cell assays with fluorescence microscopy readout
  • bottom material: #1.5 Polymer Coverslip with passivated surface (biologically inert) and RGD pattern
  • sterilized
Specifications
Specifications
Microscope Slide
2