Learn More
Ibidi μ-Slide VI 0.5 Glass Bottom
μ-Slide VI 0.5 Glass Bottom is a 6 channel slide for parallel flow assays and immunofluorescence.
Brand: Ibidi 80607
This item is not returnable.
View return policy
Description
- A 6 channel μ-Slide suitable for flow experiments and for immunofluorescence assays
- bottom material: #1.5H (170 μm ± 5 μm) D 263 M Schott glass coverslip
- sterilized
Specifications
Microscope Slide | |
15 |