Learn More
Horst™ Slim Heating Tape with High Specific Heat Output
Product Code. 10513282
Click to view available options
Length (Metric):
1.0 m
10.0 m
2.0 m
3.0 m
Wattage:
50 W
100 W
150 W
480 W
Unit Size:
Each
Product Code. 10513282
Brand: Horst™ 25207
Please to purchase this item. Need a web account? Register with us today!
Double-laid heating conductors in a slim, insulated tape.
  • Slim design.
  • Electrically insulated with PTFE.
  • Metallic protective braid of nickel-plated copper.
Specifications
Description Bending radius: 10 mm, Thickness: 4 mm
Length (Metric) 10.0 m
Voltage 230 V
Material PTFE, Nickel-plated Copper
Length (Metric) Power Cord 1 m
Wattage 480 W
Width (Metric) 4 mm
Temperature (Metric) 250°C (Nominal)