Learn More
LevGo™ SmartSlicer™ Safety Razor Blades
Click to view available options
Size:
Small
Large
Unit Size:
Pack of 100
Specifications
Specifications
| Blade Material | Food Grade PET |
| Product Type | Blade |
| For Use With (Application) | Slicing Agarose Gels, Cutting Soft Tissue and Plant Matter |
| Size | Large |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Thickness (Metric) | 0.22 mm |
| Certifications/Compliance | Food Grade |
| Autoclavable | Not Autoclavable |
| Quantity | 100 Pk. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido