missing translation for 'onlineSavingsMsg'
Learn More
Learn More

- Home
- Products
- Chemicals
- Salts and Inorganics
- Inorganic Salts
- Alkali Metal Salts
- Sodium Dichromate Dihydrate, Honeywell Fluka™
Favorite item to keep tabs on it!
Saved!
Go to My Lists
You have reached the limit of 350 favorited items.
Manage Your Favorites
Sodium Dichromate Dihydrate, Honeywell Fluka™
Product Code. 15671110 Shop All Honeywell Fluka Products1 kg
Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable.
View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15671110
Brand: Honeywell-Fluka 122661KG
This item is not returnable.
View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Description
- ≥95% for general laboratory use.

Specifications
Chemical Identifiers
CAS | 7789-12-0 |
Molecular Formula | Cr2H4Na2O9 |
Molecular Weight (g/mol) | 297.995 |
MDL Number | MFCD00149166 |
InChI Key | JYDRNIYTFCBIFC-UHFFFAOYSA-N |
Synonym | sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903Show MoreShow Less |
PubChem CID | 108005 |
IUPAC Name | disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate |
SMILES | O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+] |
Specifications
Quantity | 1 kg |
Linear Formula | Na2Cr2O7 · 2H2O |
UN Number | 3086 |
Formula Weight | 298g/mol |
Percent Purity | ≥99% |
Grade | purum |
Chemical Name or Material | Sodium Dichromate Dihydrate |
Product Suggestions
You may be interested in these alternatives
-
Savings of 78.09 €Product Code. 10216610
-
-
Product Code. 10348640
-
Product Code. 11344217
-
Product Code. 12662690
Customers who viewed this item also viewed This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
Viewing 1-4 of
-
Catalog No. 10703811
198,64 € / 500g -
Catalog No. 10793611
69,78 € / 1kg -
Catalog No. 10117113
35,95 € / 100g -
Catalog No. 10112640
17,15 € / 1kg -
Catalog No. 10092740
27,44 € / 1kg -
Catalog No. 10401841
28,04 € / 500mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Dichromate Dihydrate, Honeywell Fluka™ >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Spot an opportunity for improvement?Share a Content Correction