missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Solution 1M (1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical
Click to view available options
Quantity:
1 Ampule
Description
For volumetric analysis
Specifications
Specifications
pH | 12 |
Boiling Point | >100°C |
CAS | 1310-73-2 |
Melting Point | -10°C |
Molecular Formula | HNaO |
MDL Number | 3548 |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Quantity | 1 Ampule |
SMILES | [OH-].[Na+] |
Show More |
Safety and Handling
Product Identifier
- Sodium hydroxide 1M solution (10M concentrate ampoules)
- Danger
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Sodium hydroxide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction