Learn More
Sodium Picosulfate, MedChemExpress
MedChemExpress Sodium Picosulfate inhibits absorption of water and electrolytes, and increases their secretion.
Brand:  Medchem Express HY-B0544
Encompass_Preferred
 View more versions of this product
Product Code. 16418366
  • 201.15 € / 100mg
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
Sodium Picosulfate inhibits absorption of water and electrolytes, and increases their secretion.
Specifications
Specifications
Sodium Picosulfate
White
C18H13NNa2O8S2
Sodium Picosulphate
O=S(OC1=CC=C(C=C1)C(C2=CC=CC=N2)C3=CC=C(OS(=O)(O[Na])=O)C=C3)(O[Na])=O
481.41
Research
Metabolism-protein/nucleotide metabolism
10040-45-6
Research
100 mg
DMSO : 100 mg/mL (207.72 mM; Need ultrasonic) ∣H2O : ≥ 100 mg/mL (207.72 mM)
481.41
98.0%
Solid