Learn More
Diversey Soft Care Sensitive
Fragrance-free, dye-free mild hand wash for sensitive skins. Proves a great solution for areas where frequent washing is required and healthy hands are desired.
35.25 € - 40.15 €
Specifications
Product Type | Gentle Soap |
---|---|
Color | Clear |
Form | Liquid |
Disposable | Single-use |
Latex Free | Yes |
Product Code | Brand | Size | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Packaging | Price | Quantity & Availability | |||||
17893105
|
Diversey
101108190 |
6 x 1 L |
35.25 €
6 L |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
17853105
|
Diversey
101108201 |
2 x 5 L |
40.15 €
10 L |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
17873105
|
Diversey
101108206 |
10 x 0.5 L | With pump |
37.30 €
5 L |
Estimated Shipment: 24-04-2025
Log in to see stock. |
|||||
Description
- Biodegradable and ethically sourced
- Skin compatability tested (Patch test)
- Eco-certified with Nordic Swan Ecolabel
- Endorsed by Swedish Allergy and Asthma Foundation
- Safe, quick and effective hand wash helps reduce the spread of germs
- Effective surfactant system ensures excellent removal of grease and other soils
- Wetting agents provide even application and an easy rinse
- Reduced skin irritation and greater skin compatibility with a pH balanced formula
- Moisturizes hands with softening emollients, maintaining skinhealth for frequent users
- Rubs easily onto hands and leaves no sticky residue
Specifications
Gentle Soap | |
Liquid | |
Yes |
Clear | |
Single-use | |
Non-sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Diversey Soft Care Sensitive