Learn More
Memmert™ Software Atmocontrol, Base Licence For 1 Unit, Iq/Oq Doc. Available In De And En, For S55
Memmert™ AtmoCONTROL Software meets the requirements for the use of electronically stored data sets and electronic signatures.
Brand:  Memmert™ S55FDAQ1
Product Code. 16325927
Description
Description
  • Software conforming to FDA AtmoCONTROL
  • Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
  • Base licence for the control of one unit
  • Respective IQ/OQ documents available in German and English language (without surcharge)
Specifications
Specifications
Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
Models U, UF, TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 30, 55, 75, 110, 160, 260, 400, 450, 750, 106
Software
Product Suggestions
Product Suggestions
Videos
Videos
  • Memmert AtmoCONTROL (EN)
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Memmert AtmoCONTROL (EN)
    • Memmert ControlCOCKPIT (EN)
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
        Memmert ControlCOCKPIT (EN)
      • Memmert AtmoCONTROL (DE)
        Video Player is loading.
        Current Time 0:00
        Duration 1:36
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 1:36
        1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Memmert AtmoCONTROL (DE)
      • Memmert ControlCOCKPIT (DE)
        Video Player is loading.
        Current Time 0:00
        Duration 1:15
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 1:15
        1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Memmert ControlCOCKPIT (DE)
      SDS
      Documents
      Documents
      Certificates
      Promotions
      Promotions
      Product Content Correction
      Your input is important to us. Please complete this form to provide feedback related to the content on this product.
      Product Title
      Memmert™ Software Atmocontrol, Base Licence For 1 Unit, Iq/Oq Doc. Available In De And En, For S55 >