Learn More
Memmert™ Software AtmoCONTROL for UN160plus
AtmoCONTROL Software meets the requirements for the use of electronically stored data sets and electronic signatures.
Brand: Memmert™ UN160plusFDAQ1
This item is not returnable.
View return policy
Description
- Software conforming to FDA AtmoCONTROL
- Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
- Base licence for the control of one unit
- Respective IQ/OQ documents available in German and English language (without surcharge)
Specifications
Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA) | |
UN160plus Oven | |
Software |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Software AtmoCONTROL for UN160plus > For Use With: UN160plus
Spot an opportunity for improvement?Share a Content Correction