missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B Braun™ Solofix™ Disposable Steel Lancets
For use in capillary blood collection.
Brand: B Braun™ 6185002
This item is not returnable.
View return policy
Description
- Sterile
- Single use
- Manufactured from stainless steel
- Single lancet in sterile peel package, 5 pieces per slip, dispenser box with 500 pieces
- Gamma ray irradiation sterilization
Specifications
EN ISO 13485 | |
Capillary blood collection | |
39.1 mm | |
500/box | |
5.6 mm |
Single-use | |
2.9 mm exposed | |
Stainless steel | |
Sterile | |
Lancet |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
B Braun™ Solofix™ Disposable Steel Lancets > 500/box
Spot an opportunity for improvement?Share a Content Correction