Learn More
Ansell Edmont™ Solvex™ 37-155 Series Green Nitrile Immersion Gloves
Product Code. 15221099
This item is not returnable. View return policy
This item is not returnable. View return policy
Specifications
Back Material Nitrile
Certifications/Compliance FDA : 21 CFR 177.2600, EN 388 : 3102
Cuff Material Nitrile
Product Type Immersion Gloves
Thickness (English) 15 mil
Glove Cuff Color Green
Glove Cuff Style Straight
Palm Type Pebble
Quantity 72 Pairs
Resistant To Chemical and liquid resistance
Series 37-155