missing translation for 'onlineSavingsMsg'
Learn More

Sotorasib, MedChemExpress

Product Code. 16454808
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16454808

Brand: Medchem Express HY114277

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sotorasib (AMG-510) is a first-in-class, orally bioavailable, and selective KRAS G12C covalent inhibitor. Sotorasib irreversibly inhibits KRAS G12C by locking it in an inactive GDP-bound state. Sotorasib is the first KRAS G12C inhibitor in clinical development and leads to the regression of KRAS G12C tumors.

Sotorasib (AMG-510) is a first-in-class, orally bioavailable, and selective KRAS G12C covalent inhibitor. Sotorasib irreversibly inhibits KRAS G12C by locking it in an inactive GDP-bound state. Sotorasib is the first KRAS G12C inhibitor in clinical development and leads to the regression of KRAS G12C tumors.

Specifications

Chemical Name or Material Sotorasib
Physical Form Powder
CAS 2296729-00-3
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C30H30F2N6O3
For Use With (Application) Cancer-Kinase/protease
Synonym AMG-510
Solubility Information DMSO : 50 mg/mL (89.19 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (59.46 mM; ultrasonic and adjust pH to 11 with NaOH)
SMILES O=C(C=C)N1C[C@H](C)N(C2=NC(N(C3=C(C)C=CN=C3C(C)C)C4=C2C=C(F)C(C5=C(O)C=CC=C5F)=N4)=O)CC1
Molecular Weight (g/mol) 560.59
Formula Weight 560.59
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.