Learn More
Camping Gaz Soudogaz X2000 PZ Welder
For those looking for an excellent quality to price ratio, these welders for pierceable cartridge represent a valid alternative to those for CV valve cartridge. Once connected, the cartridge can no longer be disconnected.
Brand: Camping Gaz 2000026177
This item is not returnable.
View return policy
Description
- High performance multi-purpose blow torch with a high quality brass burner.
- Metallic cartridge cover with plastic screw-on base for easy cartridge insertion.
- To use with cartridge C 206 GLS.
Specifications
23.5 cm | |
12.5 cm | |
13.7 cm | |
0.64 kg |
Black, Blue | |
Soft Soldering and Brazing | |
1800°C |