Learn More
Bel-Art™ SP Scienceware™ Fast-Release Pipette Pump™ II Pipetters
Includes a rapid release valve for quick dispensing.
43.74 €
Specifications
Autoclavable Not Autoclavable
Material Polypropylene, Silicone Rubber
Type Fast Release Pipet Filler
View More Specs
Products 1
Product Code Brand Color For Use With (Application) Price Quantity & Availability  
Product Code Brand Color For Use With (Application) Price Quantity & Availability  
30193418
View Documents
Bel-Art™
F37911-1010
Green Pipets up to 10 mL
43.74 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description
Description
  • Rotate the knurled thumb wheel to aspirate liquid to the desired volume
  • Apply light pressure to the fast release lever to rapidly dispense
  • Rotate the thumb wheel for gradual dispensing
  • Glass or plastic pipettes are easily inserted into a unique collar with stabilizing fingers for a secure hold
  • Sizes are color coded by size for easy identification
  • Can be disassembled for cleaning
Specifications
Specifications
Not Autoclavable
Polypropylene, Silicone Rubber
Fast Release Pipet Filler
Videos
SDS
Documents
Documents
Product Certifications