Learn More
Lenz Laborglasintrumente Spare Valve Spindles
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 02280002
Description
- Spare spindles for needle-valve stopcocks
- Material: PTFE (spindle), PP (screw cap and retaining device)
- Easy to change
- Pack: 1 piece
Specifications
PTFE | |
Needle-valve stopcocks |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.