missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ SPDEF Recombinant Protein

Brand:  Abnova™ H00025803-P01.10ug

 View more versions of this product

Product Code. 16127762

  • 335.00 € / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH21299
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
62.59
8
Glutathione Sepharose 4 Fast Flow
10 ug
MGSASPGLSSVSPSHLLLPPDTVSRTGLEKAAAGAVGLERRDWSPSPPATPEQGLSAFYLSYFDMLYPEDSSWAAKAPGASSREEPPEEPEQCPVIDSQAPAGSLDLVPGGLTLEEHSLEQVQSMVVGEVLKDIETACKLLNITADPMDWSPSNVQKWLLWTEHQYRLPPMGKAFQELAGKELCAMSEEQFRQRSPLGGDVLHAHLDIWKSAAWMKERTSPGAIHYCASTSEESWTDSEVDSSCSGQPIHLWQFLKELLLKPHSYGRFIRWLNKEKGIFKIEDSAQVARLWGIRKNRPAMNYDKLSRSIRQYYKKGIIRKPDISQRLVYQFVHPI
PDEF/RP11-375E1__A.3/bA375E1.3
SPDEF
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
25803
SPDEF (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SPDEF
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions