Learn More
Tocris Bioscience™ SR 3576
Highly potent and selective JNK3 inhibitor
Brand: Tocris Bioscience™ 5044/50
580.54 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
SR 3576 is a highly potent and selective JNK3 inhibitor (IC50 = 7 nM); exhibits > 2800-fold selectivity over p38.Specifications
>97% | |
SR 3576 | |
SR 3576 |
50 mg | |
501.53 |
For Research Use Only