Learn More
SSEA5 Monoclonal Antibody (8E11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1144
290.01 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Immunofluorescence and flow cytometric analysis of MA1-144 shows positive staining of SSEA-5 positive pluripotent cells within cultured human embryonal carcinoma NCCIT and NTERA-2 cells. Mouse teratocarcinoma cell line F9 was used as a negative control and showed no staining.
Stage-Specific Embryonic Antigen-5 (SSEA-5) is an oligosaccharide that can be part of glycosphingolipids and glycoproteins. It contains the motif Fuc1-2Gal-beta1-3GlcNAc-beta, which is characteristic of H-1 antigens. SSEA-5 is found in the inner cell mass of the human blastocyst during embryogenesis, on undifferentiated human embryonic stem cells and on the surface of induced pluripotent cells. SSEA-5 was found to proceed the loss of other known stem cell markers and the combination of SSEA-5 with either CD9 and CD90 or CD50 and CD200 was superior in identifying and eliminating rare undifferentiated human ES cells from differentiated cell populations than the use of other established stem cell markers.
Specifications
SSEA5 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry, Immunocytochemistry | |
8E11 | |
Unconjugated | |
SSEA-5 | |
Undifferentiated H9 human embryonic stem cells | |
100 μg | |
Primary | |
-20°C | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.