Learn More
Karl Hecht™ Assistent™ ThermoClean™ DC Stabilizing Solution
Prevents algae/bacterial growth and stops odor formation in water baths. Karl Hecht™ Assistent™ ThermoClean™ DC Stabilizing Solution is suitable for all kinds of thermostats and circulation pumps made of metal, glass or any plastics.
Brand: Karl Hecht™ 43170001
Description
- Solvent-free
- Only minor pH-change to the water
- Prevents foam formation, even with strong circulation
- No obstructions in tubings and temperature control systems
- Can also be used for closed temperature control systems
- Free of aggressive phosphates and oxidizing or reducing substances
- Available in blue and clear variants
Specifications
Blue | |
Stabilizing Solution | |
100 mL |
Safety and Handling
- ThermoClean DC
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.