Learn More
Cole-Parmer™ Stainless Steel Balance
Brand: Cole-Parmer™ 20000-33
This item is not returnable.
View return policy
Specifications
150 g | |
0.02 g | |
220 V |
LCD | |
Stainless Steel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer™ Stainless Steel Balance > Capacity: 150g
Spot an opportunity for improvement?Share a Content Correction