Learn More
Memmert™ Stainless Steel Bottom Drip Tray for UF750, Reinforced Chamber Option
Non-perforated bottom drip trays constructed of stainless steel.
Brand:  Memmert™ UF750B34055
Product Code. 17529545
  • 380.00 € / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
Specifications
UF750 Oven
Bottom Drip Tray
Stainless Steel
Rim: 15 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Stainless Steel Bottom Drip Tray for UF750, Reinforced Chamber Option > For Use With: UF750