Learn More
BINDER™ Expansion Rack
Expansion Rack constructed of stainless steel.
486.00 € - 755.00 €
Specifications
Description | Expansion racks - stainless steel |
---|---|
Type | Rack |
Material | Stainless Steel |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
16616463
|
BINDER™
8009-1091 |
BINDER Chambers VD 115, VDL 115 | |||||||
16626463
|
BINDER™
8009-1092 |
BINDER Chambers VD 56, VDL 56 | |||||||
16636463
|
BINDER™
8009-1093 |
BINDER Chambers VD 23, VDL 23 | |||||||
Description
- Stainless steel
- Accessory rack
Specifications
Expansion racks - stainless steel | |
Rack | |
Stainless Steel |