Learn More
BINDER™ Expansion Rack, Includes Calibration
Use this stainless steel expansion rack instead of the standard aluminum expansion rack. Includes calibration.
600.00 € - 840.00 €
Specifications
Description | Expansion racks - stainless steel expansion rack instead of standard aluminum expansion rack, incl. calibration |
---|---|
Type | Rack |
Includes | Calibration |
Material | Stainless Steel |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
16616483
|
BINDER™
8612-0011 |
BINDER Chambers VD 23, VDL 23 |
600.00 €
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
16626483
|
BINDER™
8612-0012 |
BINDER Chambers VD 56, VDL 56 |
720.00 €
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
16636483
|
BINDER™
8612-0013 |
BINDER Chambers VD 115, VDL 115 |
840.00 €
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
Description
- Stainless steel
- Accessory rack
Specifications
Expansion racks - stainless steel expansion rack instead of standard aluminum expansion rack, incl. calibration | |
Rack | |
Calibration | |
Stainless Steel |