Learn More
Blanco™ Gastronorm Stainless Steel Containers
Made in Germany in a range of sizes and depths. Blanco™ Gastronorm Stainless Steel Containers are dishwasher-safe, stackable containers made of AISI 304 stainless steel. They can be used for a variety of applications at a wide range of temperatures, from food service/catering to goods storage.
38.00 € - 151.00 €
Specifications
Product Type Gastronorm Container
Material Stainless Steel (304)
Certifications/Compliance European standard EN 631
Quantity 1 Each
Temperature Range (Metric) −40°C to 280°C
View More Specs
Products 10
Product Code Brand Size Depth (Metric) Volume (Metric) Price Quantity & Availability  
Product Code Brand Size Depth (Metric) Volume (Metric) Price Quantity & Availability  
11933836
View accessory products View Documents
Blanco™
1550575
GN 1/4 150 mm 4 L
76.00 €
Each
Estimated Shipment: 22-07-2025
to see stock.
Add to basket
11953826
View accessory products View Documents
Blanco™
1550583
GN 1/2 150 mm 8.9 L
57.00 €
Each
Estimated Shipment: 05-05-2025
to see stock.
Add to basket
11943826
View accessory products View Documents
Blanco™
1550582
GN 1/2 150 mm 6 L
41.00 €
Each
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
11973816
View Documents
Blanco™
1550586
GN 2/3 100 mm 8.5 L
58.00 €
Each
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
11993826
View Documents
Blanco™
1550594
GN 2/1 100 mm 28.9 L
151.00 €
Each
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
11938017
View accessory products View Documents
Blanco™
1550579
GN 1/3 150 mm 5.5 L
59.00 €
Each
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
11948017
View accessory products View Documents
Blanco™
1550578
GN 1/3 100 mm 3.8 L
38.00 €
Each
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
11903826
View accessory products View Documents
Blanco™
1550592
GN 1/1 200 mm 26.5 L
129.00 €
Each
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
11983816
View accessory products View Documents
Blanco™
1550590
GN 1/1 100 mm 13.3 L
54.00 €
Each
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
11923836
View accessory products View Documents
Blanco™
1550574
GN 1/4 100 mm 2.7 L
39.00 €
Each
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
Description
Description
Gastronorm Stainless Steel Containers feature outstanding stability thanks to circumferential edge stamping. These rugged and dimensionally stable containers are easy to clean and offer optimum hygiene thanks to the smooth, non-porous surface.
  • First-class, corrosion-resistant finish with burr-free edges
  • Temperature-resistant from −40°C to +280°C
  • Constructed of tasteless and odorless stainless steel (AISI 304)
  • Exact corner finishing and stacking collars prevent jamming and wedging during stacking
Gastronorm containers can be stacked and combined in a variety of combinations. Standards sizes include:
  • GN 2/1: 650mmL x 530mmH
  • GN 1/1: 530mmL x 325mmH
  • GN 1/2: 325mmL x 265mmH
  • GN 1/4: 265mmL x 162mmW
  • GN 2/3: 354mmL x 325mmH
  • GN 1/3: 325mmL x 176mmH
  • GN 1/6: 176mmL X 162mmH
Specifications
Specifications
Gastronorm Container
European standard EN 631
−40°C to 280°C
Stainless Steel (304)
1 Each
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Blanco™ Gastronorm Stainless Steel Containers