Learn More
Bandelin Electronic™ Sonorex™ Insert Baskets for Ultrasonic Baths
Facilitates ultrasonic applications and simultaneously protects oscillating tank and parts to be cleaned. Bandelin Electronic™ Sonorex™ Insert Baskets for Ultrasonic Baths are manufactured from stainless steel.
Brand:  Bandelin Electronic™ 205
 View more versions of this product
Product Code. 10384581
  • 277.00 € / Each
Estimated Shipment: 22-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Facilitates ultrasonic applications and simultaneously protects oscillating tank and parts to be cleaned. Bandelin Electronic™ Sonorex™ Insert Baskets for Ultrasonic Baths are manufactured from stainless steel.
  • Available for a wide range of tank capacities
  • Available in a range of mesh sizes
  • Excellent for the cleaning of technical glassware, analysis sieves, medical instruments, metal parts, electronic components, degassing, emulsifying and sample preparation
Specifications
Specifications
Basket Insert
275 mm
Stainless Steel
245 x 50 x 275 mm
Ultrasonic Bath, RK 514 BH, DK 514 BP
245 mm
50 mm
K14B