Learn More
Welch™ Stainless Steel KF Inner Centering with Buna N O-ring
Shop All Welch Ilmvac Products
Click to view available options
Diameter (Metric) Outside:
16 mm
25 mm
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) Outside | 25 mm |
| For Use With | Separator 25 DN |
| Material | Stainless Steel |
| Quantity | 1 Each |
| Type | KF Inner Centering |