Learn More
Bochem™ Stainless Steel Rods
Product Code. p-8005461
Click to view available options
Length (Metric):
500 mm
600 mm
750 mm
1000 mm
1500 mm
Length (Metric) Rod:
500 mm
600 mm
750 mm
1000 mm
1500 mm
Unit Size:
Each
Product Code. 10509861
Brand: Bochem™ 5123
Please to purchase this item. Need a web account? Register with us today!
Use Bochem™ Stainless Steel Rods for strength for your lab operations.
  • Stainless Steel
  • Sturdy
Specifications
Material Stainless Steel
Length (Metric) 1000 mm
Length (Metric) Rod 1000 mm
Diameter (Metric) Rod 12 dia.
For Use With (Application) Stand Bases