Learn More
Sampling Systems™ Stainless Steel Sample Splitter Riffle Boxes
Product Code. 15809541 Shop All Sampling Systems Products
Click to view available options
Width (Metric) Chute:
6 mm
10 mm
20 mm
30 mm
No. of Chutes:
12
16
Unit Size:
Each
Product Code. 15809541
Brand: Sampling Systems™ 8601S30
Only null left
Add to basket
Add to basket
These easy-to-use Sample Splitters in stainless steel are for dividing and reducing bulk samples.
  • Simply drop the sample into the top and the sample is split in two
  • Split sample drops into the riffle boxes underneath
  • It is recommended that width of slots are 2.5 to 3 times bigger than the diameter of the largest particle being split
  • Available in a wide range of sizes
Specifications
Product Type Riffle Box
Material Stainless Steel
No. of Chutes 12
For Use With (Application) Dividing and reducing bulk samples
Width (Metric) Chute 30 mm