Learn More
Rogo Sampaic™ Stainless Steel Scissors
Excellent for a variety of cutting applications in the laboratory. Rogo Sampaic™ Stainless Steel Scissors have a beautiful polished matte finish.
47.00 € - 87.00 €
Product Code | Brand | Tip Style | For Use With (Application) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tip Style | For Use With (Application) | Length (Metric) | Price | Quantity & Availability | |||
11738531
|
Rogo Sampaic™
FMN015 |
Round/Round | Anatomical | 140 mm |
87.00 €
Pack of 10 |
Estimated Shipment: 10-12-2024
Log in to see stock. |
|||
11706145
|
Rogo Sampaic™
FMN025 |
Sharp/Sharp | Noyes micro | 115 mm |
47.00 €
Each |
In Stock
Log in to see stock. |
|||
Description
- Surgical grade stainless steel
- Available in a variety of styles - Metzenbaum, dissecting, iris, anatomical
Specifications
Stainless Steel |