Learn More
De Buyer™ Stainless Steel Serving Dishes
Click to view available options
Volume (Metric):
2.6 L
3.6 L
4.9 L
Unit Size:
Each
Specifications
Specifications
| Product Type | Dish |
| Material | Stainless Steel |
| Height (Metric) | 45 mm |
| Length (Metric) | 350 mm |
| Quantity | Each |
| Shape | Rectangular |
| Volume (Metric) | 3.6 L |
| Width (Metric) | 270 mm |