Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for UN260plus, 15 mm
Non-perforated slide-in drip trays constructed of stainless steel.
Brand:  Memmert™ UN260plusB29726
Product Code. 17590016
  • 190.00 € / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
Specifications
UN260plus Oven
Drip Tray
Stainless Steel
Rim: 15 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for UN260plus, 15 mm > For Use With: UN260plus