Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for UN260plus, 15 mm
Non-perforated slide-in drip trays constructed of stainless steel.
Brand: Memmert™ UN260plusB29726
This item is not returnable.
View return policy
Description
- Accessory
- May affect temperature distribution
- Constructed of stainless steel
- Rim height: 15 mm
Specifications
UN260plus Oven | |
Drip Tray | |
Stainless Steel | |
Rim: 15 mm |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for UN260plus, 15 mm > For Use With: UN260plus
Spot an opportunity for improvement?Share a Content Correction