Learn More
Memmert™ Stainless Steel Slide-In Drip Tray
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP260eco
IPP410eco
IPP750eco
UN110pa
For Use With (Application):
Do not use with Modified Chamber
Reinforced Chamber Option K1
Unit Size:
Each
Description
- May affect the temperature distribution
- 15 mm rim
Specifications
Specifications
| Description | Stainless steel slide-in drip tray, 15 mm rim, with guide bars and fixing screws (may affect the temperature distribution) - can be used only in connection with option K1 |
| For Use With (Equipment) | IPP750eco |
| Type | Drip Tray |
| For Use With (Application) | Reinforced Chamber Option K1 |
| Length (Metric) | 15 mm rim |