Learn More
Memmert™ Stainless Steel Slide-In Drip Tray
Product Code. 17150265 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP260eco
IPP410eco
IPP750eco
UN110pa
For Use With (Application):
Do not use with Modified Chamber
Reinforced Chamber Option K1
Unit Size:
Each
Product Code. 17150265
Brand: Memmert™ IPP750ecoB02075
Only null left
Add to basket
Add to basket
Stainless steel slide-in bottom drip tray
  • May affect the temperature distribution
  • 15 mm rim
Specifications
Description Stainless steel slide-in drip tray, 15 mm rim (may affect the temperature distribution) - cannot be used in connection with option K1
For Use With (Equipment) IPP750eco
Type Drip Tray
For Use With (Application) Do not use with Modified Chamber
Length (Metric) 15 mm rim