Learn More
Bode Chemie™ Stellisept Med™ Antimicrobial Washing Lotion
Product Code. 10689213
Product Code. 10689213
Brand: Bode Chemie™ 9805012
Only null left
Add to basket
Add to basket
Use as a skin friendly alternative to conventional decontamination preparations. Bode Chemie™ Stellisept Med™ Antimicrobial Washing Lotion is suitable for antimicrobial body cleansing and MRSA eradication in accordance with the Robert Koch-Institute recommendation.
  • Mild antimicrobial rinse-off wash lotion
  • Comprehensive activity against bacteria within 30 seconds incl. MRSA/ ORSA and VRE
  • Pleasant scent
  • No alcohol and colorants
Specifications
Form Liquid
pH 5.5 to 6.5
Appearance Clear
Composition Aqua, PEG–40 hydrogenated castor oil, PEG-150 distearate, glycerin, panthenol, PEG-7 glyceryl
Container Type Bottle