Learn More
Sampling Systems™ SteriWare™ Micro-Spatulas
BIOPHARMA
Product Code. 15809401 Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
0.3 mL
0.5 mL
2 mL
Color:
Red
White
Material:
HDPE
Polystyrene
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Product Code. 15809401
Brand: Sampling Systems™ 8015V170S
Please to purchase this item. Need a web account? Register with us today!
Allows samples to be taken from very small containers such as vials with its narrow sampling blade. Sampling Systems™ SteriWare™ Micro-Spatulas are ideal for handling small quantities of powders and granules.
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Different blade widths available.  Ideal for small vials or sample containers
  • Also available in HDPE for extra chemical resistance
Specifications
Sterility Sterile
Capacity (Metric) 0.3 mL
Color White
Disposable Single-use
Width (Metric) 4 mm
Material Polystyrene
Length (Metric) 170 mm