missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ibidi Sticky-Slide VI
Click to view available options
:
Height: 400 μm
Description
- A 6 channel slide suitable for flow experiments and for immunofluorescence assays
- bottomless with self-adhesive underside
- sterilized
Specifications
Specifications
| Length (Metric) | 400 μm |
| Sterility | Sterile |
| Type | Microscope Slide |
| Length (English) | 0.02 in. |
| Quantity | 15 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction