Learn More
Usbeck™ Straight Blunt Point Nickle Plated Forceps
Dissect specimens with accuracy. Usbeck™ Straight Blunt Point Nickle Plated Forceps are perfect for use in dissection or handling lab specimens.
3.10 € - 3.60 €
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10065571
|
Usbeck™
3000 |
105 mm |
3.10 €
Each |
Estimated Shipment: 25-03-2025
Log in to see stock. |
|||||
10065581
|
Usbeck™
3003 |
145 mm |
3.60 €
Each |
Estimated Shipment: 25-03-2025
Log in to see stock. |
|||||
Description
- Straight
- Blunt points
Specifications
Nickel-plated Steel |