Learn More
Techne™ Stuart™ 96-well Low Profile PCR Plates
Product Code. 13088523
Click to view available options
Color:
Black
Clear
White
Unit Size:
Pack of 25
Product Code. 13088523
Brand: Techne™ 1621033
Please to purchase this item. Need a web account? Register with us today!
Stuart™ 96-well Low Profile PCR Plates have shorter wells than the standard profile plates, decreasing the dead space between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction.
  • Low profile design decreases dead space between the lid of the thermal cycler, increasing PCR efficiency
  • Available in Black, White and Clear
  • Non-sterile
Specifications
Format Standard
Array 8 x 12
Color Black
No. of Wells 96
Sterility Non-sterile
No. per Pack 25
Profile Low
Quantity 25 Pk.