Learn More
Techne™ Non-skirted 96-well PCR Plates
Product Code. 13008533
Click to view available options
Color:
Clear
Unit Size:
Pack of 25
Product Code. 13008533
Brand: Techne™ 9996789
Please to purchase this item. Need a web account? Register with us today!
These non-skirted PCR plates fit the blocks of most thermal cyclers and real-type PCR instruments, they are not suitable for robotic use.
  • Low profile
  • Fits most blocks of thermal cyclers
  • Not suitable for robotic instruments
Specifications
Format Standard
Array 8 x 12
Color Clear
Product Type 96-well PCR Plate
No. of Wells 96
No. per Pack 25
Profile Low
Quantity 25 Pk.